Order Entry
Export
ContactUsLinkComponent
Anti-OGT Rabbit Polyclonal Antibody
Anti-OGT Rabbit Polyclonal Antibody
  BSBTPB9767
undefined
Anti-OGT Rabbit Polyclonal Antibody
  BSBTPB9767

 

  • Antibody type:
    Primary
  • Antigen symbol:
    OGT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    8473
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit(OGT) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: OGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat