Safety can, oily waste
These oily waste cans with galvanised steel body construction with durable powder-coat finish are essential when using solvent soaked cloths and wiping rags. Rags and cloths soaked with thinners, linseed oil, combustible adhesives and other flammable liquids present a serious fire risk when discarded improperly.
- Available in several sizes and various colours
- Lid opens no more than 60° and stays closed when not in use which isolates the contents from fire sources and limits oxygen, virtually eliminating a risk of spontaneous combustion
Compliance with OSHA and FM approved.