Order Entry
Export
ContactUsLinkComponent
Anti-KLHL36 Rabbit Polyclonal Antibody
Anti-KLHL36 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9628R
Supplier:  Bioss
Anti-KLHL36 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9628R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Kelch-like 36
  • Antigen symbol:
    KLHL36
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    Kelch like protein 36|Kelch like 36|KLH36_HUMAN|kelch-like 36 Drosophila|KLHL-36.|KLHL 36|Kelch-like protein 36
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

C16orf44 is a 616 amino acid protein that contains six Kelch repeats, one BTB/POZ domain and one BTB/Kelch associated (BACK) domain. C16orf44 is believed to play a role in protein ubiquitination and may function as a substrate-specific adapter of an E3 ubiquitin-protein ligase complex. E3 ligases accept a ubiquitin residue from an E2 ubiquitin-conjugating enzyme and immediately transfer that residue to a protein that is targeted for degradation. Specifically, C16orf44 interacts with CUL-3, a member of the cullin family of mediators that participate in the selective targeting of proteins for ubiquitin-mediated proteolysis. Due to alternative splicing events, two isoforms of C16orf44 are expressed.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: KLHL36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat