Specifications
- Antibody type:Primary
- Antigen symbol:CLLD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:SPRY7_HUMAN.|Chromosome 13 open reading frame 1|CLL deletion region gene 6 protein|Chronic lymphocytic leukemia deletion region gene 6 protein|Hypothetical protein LOC57213|CLLD6|CLLD 6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
CLLD6, is a 196 amino acid protein that contains one B30.2/SPRY domain. Expression of CLLD6 is found at highest levels in heart, skeletal muscle and testis, with lower levels found in thymus, peripheral blood leukocytes, lymph node and bone marrow. Existing as two alternatively spliced isoforms, CLLD6 maps to human chromosome 13, which comprises nearly 4% of human DNA and contains about 114 million base pairs and 400 genes. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. RB1 encodes a crucial tumor suppressor protein which, when defective, leads to malignant growth in the retina and has been implicated in a variety of other cancers. The gene SLITRK1, which is associated with Tourette syndrome, is on chromosome 13. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CLLD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: