Order Entry
Export
ContactUsLinkComponent
Anti-ALG11 Rabbit Polyclonal Antibody
Anti-ALG11 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7957R
Supplier:  Bioss
Anti-ALG11 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7957R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Asparagine-linked glycosylation 11
  • Antigen symbol:
    ALG11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    440138
  • Antigen synonyms:
    alg11|AI849156|GT8|alpha-1,2-mannosyltransferase homolog yeast|Asparagine-linked glycosylation 11|ALG11_HUMAN|Asparagine-linked glycosylation protein 11 homolog|UTP14C.
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mannosyltransferase involved in the last steps of the synthesis of Man5GlcNAc(2)-PP-dolichol core oligosaccharide on the cytoplasmic face of the endoplasmic reticulum. Catalyzes the addition of the 4th and 5th mannose residues to the dolichol-linked oligosaccharide chain.Involvement in disease:Defects in ALG11 are the cause of congenital disorder of glycosylation type 1P (CDG1P). A multisystem disorder caused by a defect in glycoprotein biosynthesis and characterized by under-glycosylated serum glycoproteins. Congenital disorders of glycosylation result in a wide variety of clinical features, such as defects in the nervous system development, psychomotor retardation, dysmorphic features, hypotonia, coagulation disorders, and immunodeficiency. The broad spectrum of features reflects the critical role of N-glycoproteins during embryonic development, differentiation, and maintenance of cell functions.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ALG11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat