Order Entry
Export
ContactUsLinkComponent
Anti-DSCC1 Rabbit Polyclonal Antibody
Anti-DSCC1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7720R
Supplier:  Bioss
Anti-DSCC1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7720R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DNA replication and sister chromatid cohesion 1
  • Antigen symbol:
    DSCC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    79075
  • Antigen synonyms:
    Defective in sister chromatid cohesion protein 1 homolog|MGC5528|Sister chromatid cohesion protein DCC1.|DCC1_HUMAN|hDCC1|DSCC1|DCC1|Defective in sister chromatid cohesion 1 homolog
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Loads PCNA onto primed templates regulating velocity, spacing and restart activity of replication forks. May couple DNA replication to sister chromatid cohesion through regulation of the acetylation of the cohesin subunit SMC3.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DSCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat