Anti-FOXD4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7127R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box D4
- Antigen symbol:FOXD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:Forkhead box protein D4-like 2|FOXD4-like 3|Forkhead box protein D4-like 4|FOXD4-like 5|Forkhead box protein D4-like 5|FOXD4-like 4|Forkhead box protein D4-like 6|Forkhead box protein D4-like 3|FOXD4-like 6|FOXD4-like 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Contains 1 fork-head DNA-binding domain.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: FOXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human