Anti-E4B Rabbit Polyclonal Antibody
100 μg
Catalog # BOSSBS-5346R
Supplier: Bioss
Jump to:
Product Details & Documents
eIF4B is required for the binding of mRNA to ribosomes. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F (referenced from swissprot).
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: E4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
bs-5346R
Datasheet
Specifications
- Size:100 μg
- Western blot:Yes
- Clonality:Polyclonal
- Modification:Ser504
- Purification:Purified by Protein A.
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Host:Rabbit
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Antigen name:Eukaryotic translation initiation factor 4B
- Cat. No.:BOSSBS-5346R
- Reactivity:Human
- Gene ID:1975
- Antigen synonyms:E 4B|4B_HUMAN.|E-4B|Eukaryotic initiation factor 4B
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:EIF4B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.



