Order Entry
Export
ContactUsLinkComponent
Anti-HTR3E Rabbit Polyclonal Antibody
Anti-HTR3E Rabbit Polyclonal Antibody
Catalog # BOSSBS-5328R
Supplier:  Bioss
Anti-HTR3E Rabbit Polyclonal Antibody
Catalog # BOSSBS-5328R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5-hydroxytryptamine (serotonin) receptor 3E, ionotropic
  • Antigen symbol:
    HTR3E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    285242
  • Antigen synonyms:
    5-HT3c1|5-HT3E|5-HT3-E
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

HTR3C belongs to the ligand-gated ion channel receptor superfamily. HTR3E is a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HTR3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human