Order Entry
Export
ContactUsLinkComponent
Anti-CYP46 Rabbit Polyclonal Antibody
Anti-CYP46 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5046R
Supplier:  Bioss
Anti-CYP46 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5046R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome P450, Family 46, Subfamily A, Polypeptide 1
  • Antigen symbol:
    CYP46
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    10858
  • Antigen synonyms:
    Cholesterol 24-hydroxylase|CP46A_HUMAN|CYP46|cytochrome P450 family 46.|CP46
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

The Cyp46 gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is expressed exclusively in the brain, where it converts cholesterol to 24S-hydroxycholesterol by adding a hydroxyl group to cholesterol, producing a product that is more soluble than cholesterol and able to be exported from the brain.Cyp46 is also known as 24S-cholesterol hydroxylase.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CYP46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: