Order Entry
Export
ContactUsLinkComponent
Anti-5HT4R Rabbit Polyclonal Antibody
Anti-5HT4R Rabbit Polyclonal Antibody
Catalog # BOSSBS-2127R
Supplier:  Bioss
undefined
Anti-5HT4R Rabbit Polyclonal Antibody
Catalog # BOSSBS-2127R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5-Hydroxytryptamine receptor 4
  • Antigen symbol:
    5HT4R
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    3360
  • Antigen synonyms:
    5-hydroxytryptamine receptor 4|HTR4|5-HT4R|5-HT4|Serotonin receptor 4|5-HT-4
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    41-58/388
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: 5HT4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat