Anti-ZFP57 Rabbit Polyclonal Antibody
Catalog # BOSSBS-12252R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 57
- Antigen symbol:ZFP57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:ZNF57_HUMAN.|bA145L22.2|TNDM1|C6orf40|ZNF698|bA145L22|Zinc Finger Protein 57
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
ZFP57 is a 452 amino acid protein that contains one KRAB domain and seven C2H2-type zinc fingers and is a member of the Krüppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZFP57 functions as a transcriptional repressor that inhibits the expression of Schwann cell-specific proteins, thereby playing a role in the development of the peripheral nervous system. ZFP57 exists as two isoforms that are produced from alternative splicing events.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ZFP57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human