Anti-MAOA + MAOB Rabbit Polyclonal Antibody
Catalog # BOSSBS-11890R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Monoamine oxidase A+B
- Antigen symbol:MAOA + MAOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:Amine oxidase flavin containing|Monoamine oxidase B|MAOB|Monoamine oxidase|AOFB_HUMAN|Adrenalin oxidase|MAO A|Amine oxidase flavin containing A|MAO|Tyramine oxidase.|AOFA_HUMAN|Amine oxidase flavin containing B|Monoamine oxidase type B|MAOA|Brunner syndrome|Monoamine oxidase type A|MAO platelet|Monoamine oxidase A|MAO brain
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Monoamine oxidase A and B catalyzes the oxidative deamination of biogenic and xenobiotic amines and has important functions in the metabolism of neuroactive and vasoactive amines in the central nervous system and peripheral tissues. MAOA preferentially oxidizes biogenic amines such as 5-hydroxytryptamine (5-HT), norepinephrine and epinephrine. MAOB preferentially degrades benzylamine and phenylethylamine
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MAOA + MAOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat