- Antibody type:Primary
- Antigen name:Glutamate transporter EAAT4 associated protein 48
- Antigen symbol:PDZ RHOGEF/GTRAP48
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:DKFZp667F1223|ARHGB_HUMAN|PDZ RhoGEF|RhoA specic guanine nucleotide exchange factor|PDZ-RhoGEF|Rho guanine nucleotide exchange factor 11|RhoGEF glutamate transport modulator.|ARHGEF11|GTRAP48|Glutamate transporter EAAT4 associated protein 48|GTRAP 48|Rho guanine exchange factor GEF 11|ARHGEF 11|Rho guanine nucleotide exchange factor GEF 11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. A similar protein in rat interacts with glutamate transporter EAAT4 and modulates its glutamate transport activity. Expression of the rat protein induces the reorganization of the actin cytoskeleton and its overexpression induces the formation of membrane ruffling and filopodia. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq, Jul 2008].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PDZ RHOGEF/GTRAP48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat