Anti-E1 glycoprotein Rabbit Polyclonal Antibody
Catalog # BOSSBS-0319R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:E1 glycoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Form:liquid
- Antigen synonyms:E1|Glycoprotein E1|Spike glycoprotein E1|E1 envelope glycoprotein|POLS_RUBVM.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: E1 glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus