Anti-PAK7 Rabbit Polyclonal Antibody
Catalog # BSBTPB9733
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:p21 protein (Cdc42/Rac)-activated kinase 7
- Antigen symbol:PAK7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57144
- Antigen synonyms:PAK5
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase PAK 7(PAK7) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: PAK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat