Tubing cutter
Catalog # REST20192
Supplier: Restek
Specifications
- Description:Tubing cutter, for 1/16" O.Ø tubing
- Seq:01
- Pk:1
Specifications
About this item
Designed to cut hard or soft copper, aluminium, brass, stainless steel, Monel® alloy, or titanium.
- Produces square, smooth cuts in ¹/₁₆" tubing
- Eliminates tubing distortion