Order Entry
Export
Orders LinkContactUsLinkComponent
 

Anti-POLA2 Mouse Polyclonal Antibody

50 μg
  ABNOH00023649-B01P
 :   Abnova
Anti-POLA2 Mouse Polyclonal Antibody
  ABNOH00023649-B01P
 :  Abnova
Anti-POLA2 Mouse Polyclonal Antibody

50 μg

 
418,00 €
   
1 * 50 µG
 

Mouse polyclonal antibody raised against a full-length human POLA2 protein.

Type: Primary
Antigen: POLA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

 
  • Size:
    50 μg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Antibody type:
    Primary
  • Epitope:
    Full length
  • ImmunoFluorescence:
    Yes
  • Amino acid number:
    1 to 598
  • Host:
    Mouse
  • Antigen name:
    Polymerase (DNA directed), alpha 2, accessory subunit
  • Cat. No.:
    ABNOH00023649-B01P
  • Reactivity:
    Human
  • Gene ID:
    23649
  • Immunogen:
    POLA2 (NP_002680.2, 1 a.a. ~ 598 a.a) full-length human protein.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    POLA2