Anti-TMEM93 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29482
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ER membrane protein complex subunit 6
- Antigen symbol:EMC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:83460
- Antigen synonyms:TMEM93
- Sequence:VVAKREGPPFISEAAVRGNAAVLDYCRTS
- Immunogen:Recombinant protein corresponding to amino acids of human TMEM93.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
TMEM93 polyclonal antibody raised against recombinant human TMEM93.
Type: Primary
Antigen: TMEM93
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human