Anti-CGNL1 Rabbit Polyclonal Antibody
ABNOPAB29471
: Abnova
- Antibody type:Primary
- Antigen name:cingulin-like 1
- Antigen symbol:CGNL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:84952
- Antigen synonyms:JACOP
- Sequence:NEKVEENSTLQQRLEESEGELRKNLEELFQVKMEREQHQTEIRDLQDQLSEMHDELDSAKRSEDREKGALIEELLQAKQDLQDLLIAK
- Immunogen:Recombinant protein corresponding to human CGNL1.
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant human CGNL1.
Type: Primary
Antigen: CGNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human