Order Entry
Export
ContactUsLinkComponent
Anti-IPO5 Rabbit Polyclonal Antibody
Anti-IPO5 Rabbit Polyclonal Antibody
  ABNOPAB29468
 :  Abnova
Anti-IPO5 Rabbit Polyclonal Antibody
  ABNOPAB29468
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    importin 5
  • Antigen symbol:
    IPO5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    3843
  • Antigen synonyms:
    imp5|RANBP5|IMB3|KPNB3|Pse1
  • Sequence:
    MPEDQVGKLEATENTISAMA
  • Immunogen:
    Recombinant protein corresponding to human IPO5.
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against recombinant human IPO5.

Type: Primary
Antigen: IPO5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human