Anti-C1orf57 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29457
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:nucleoside-triphosphatase, cancer-related
- Antigen symbol:NTPCR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:84284
- Antigen synonyms:C1orf57|HCR-NTPase
- Sequence:PVDGFYTEEVRQGGRRIGFDVVTLSGTRGPLSRVGLEPPPGKRECRVGQYVVDLTSFEQLALPV
- Immunogen:Recombinant protein corresponding to human C1orf57.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant human C1orf57.
Type: Primary
Antigen: C1orf57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human