Anti-RCOR1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29456
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:REST corepressor 1
- Antigen symbol:RCOR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:23186
- Antigen synonyms:COREST|RCOR
- Sequence:SSDEEHGGGGMRVGPQYQAVVPDFDPAKLARRSQERDNLGMLV
- Immunogen:Recombinant protein corresponding to human RCOR1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant human RCOR1.
Type: Primary
Antigen: RCOR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human