- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 17
- Antigen symbol:TNFRSF17
- Clonality:Monoclonal
- Clone:8H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a lambda
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:608
- Antigen synonyms:TNFRSF13A|CD269|BCMA|BCM
- Amino acid number:5 to 50
- Sequence:AGQCSQNEYFDSLLHACIPCQLRCSSNTPPLTCQRYCNASVTNSVK
- Immunogen:TNFRSF17 (AAH58291.1, 5 a.a. ~ 50 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against a partial recombinant TNFRSF17.
Type: Primary
Antigen: TNFRSF17
Clonality: Monoclonal
Clone: 8H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human