Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 1G11]
ABNOH00023495-M03
: Abnova
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 13B
- Antigen symbol:TNFRSF13B
- Clonality:Monoclonal
- Clone:1G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:23495
- Antigen synonyms:CD267|CVID|RYZN|TACI|IGAD2|CVID2|TNFRSF14B
- Amino acid number:2 to 165
- Sequence:SGLGRSRRGGRSRVDQEERFPQGLWTGVAMRSCPEEQYWDPLLGTCMSCKTICNHQSQRTCAAFCRSLSCRKEQGKFYDHLLRDCISCASICGQHPKQCAYFCENKLRSPVNLPPELRRQRSGEVENNSDNSGRYQGLEHRGSEASPALPGLKLSADQVALVYS
- Immunogen:TNFRSF13B (NP_036584.1, 2 a.a. ~ 165 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against a partial recombinant TNFRSF13B.
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 1G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human