Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 6D12]
ABNOH00008792-M43
: Abnova
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 11a, NFKB activator
- Antigen symbol:TNFRSF11A
- Clonality:Monoclonal
- Clone:6D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:8792
- Antigen synonyms:ODFR|RANK|OFE|OSTS|FEO|LOH18CR1|OPTB7|CD265|PDB2|TRANCER
- Amino acid number:69 to 152
- Sequence:IAPPLPCGPDEYLDSWNEEDKCLLHKVCDTGKALVAVVAGNSTTPRRCACTAGYHWSQDCECCRRNTECAPGLGAQHPLQLNKDTVCK
- Immunogen:TNFRSF11A (NP_003830.1, 69 a.a. ~ 152 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A.
Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 6D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human