Order Entry
Export
ContactUsLinkComponent
Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: 4A7]
Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00008784-M12
Supplier:  Abnova
Anti-TNFRSF18 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00008784-M12
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tumor necrosis factor receptor superfamily, member 18
  • Antigen symbol:
    TNFRSF18
  • Clonality:
    Monoclonal
  • Clone:
    4A7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    8784
  • Antigen synonyms:
    CD357|GITR|AITR|GITR-D
  • Amino acid number:
    26 to 161
  • Sequence:
    QRPTGGPGCGPGRLLLGTGTDARCCRVHTTRCCRDYPGEECCSEWDCMCVQPEFHCGDPCCTTCRHHPCPPGQGVQSQGKFSFGFQCIDCASGTFSGGHEGHCKPWTDCTQFGFLTVFPGNKTHNAVCVPGSPPAE
  • Immunogen:
    TNFRSF18 (NP_004186.1, 26 a.a. ~ 161 a.a) partial recombinant protein with GST tag.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against a partial recombinant TNFRSF18.

Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human