Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 5G5]
Catalog # ABNOH00027242-M59
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 21
- Antigen symbol:TNFRSF21
- Clonality:Monoclonal
- Clone:5G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:27242
- Antigen synonyms:DR6|CD358|BM-018
- Amino acid number:89 to 170
- Sequence:SSCPVGTFTRHENGIEKCHDCSQPCPWPMIEKLPCAALTDRECTCPPGMFQSNATCAPHTVCPVGWGVRKKGTETEDVRCKQ
- Immunogen:TNFRSF21 (AAH17730.1, 89 a.a. ~ 170 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant TNFRSF21.
Type: Primary
Antigen: TNFRSF21
Clonality: Monoclonal
Clone: 5G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human