Anti-LTBR Mouse Monoclonal Antibody [clone: 5H8]
Catalog # ABNOH00004055-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:lymphotoxin beta receptor (TNFR superfamily, member 3)
- Antigen symbol:LTBR
- Clonality:Monoclonal
- Clone:5H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:4055
- Antigen synonyms:CD18|TNFR2-RP|TNFR-RP|TNFR3|TNF-R-III|TNFCR|LT-BETA-R|TNFRSF3|D12S370
- Amino acid number:37 to 225
- Sequence:ASENQTCRDQEKEYYEPQHRICCSRCPPGTYVSAKCSRIRDTVCATCAENSYNEHWNYLTICQLCRPCDPVMGLEEIAPCTSKRKTQCRCQPGMFCAAWALECTHCELLSDCPPGTEAELKDEVGKGNNHCVPCKAGHFQNTSSPSARCQPHTRCENQGLVEAAPGTAQSDTTCKNPLEPLPPEMSGTM
- Immunogen:LTBR (NP_002333.1, 37 a.a. ~ 225 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant LTBR.
Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: 5H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human