Order Entry
Export
ContactUsLinkComponent
Anti-CD80 Mouse Monoclonal Antibody [clone: 1D7]
Anti-CD80 Mouse Monoclonal Antibody [clone: 1D7]
  ABNOH00000941-M28
 :  Abnova
Anti-CD80 Mouse Monoclonal Antibody [clone: 1D7]
  ABNOH00000941-M28
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    CD80 molecule
  • Antigen symbol:
    CD80
  • Clonality:
    Monoclonal
  • Clone:
    1D7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    941
  • Antigen synonyms:
    B7-1|B7.1|CD28LG|CD28LG1|LAB7|BB1|B7
  • Amino acid number:
    35 to 242
  • Sequence:
    VIHVTKEVKEVATLSCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDITNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPTPSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETELYAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDN
  • Immunogen:
    CD80 (NP_005182.1, 35 a.a. ~ 242 a.a) partial recombinant protein with GST tag.
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against a partial recombinant CD80.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human