Anti-PDCD1LG2 Mouse Monoclonal Antibody [clone: 5C9]
Catalog # ABNOH00080380-M28
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Programmed cell death 1 ligand 2
- Antigen symbol:PDCD1LG2
- Clonality:Monoclonal
- Clone:5C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:80380
- Antigen synonyms:PDCD1L2|CD273|PD-L2|Btdc|B7DC|PDL2|bA574F11.2
- Amino acid number:19 to 220
- Sequence:ALFTVTVPKELYIIEHGSNVTLECNFDTGSHVNLGAITASLQKVENDTSPHRERATLLEEQLPLGKASFHIPQVQVRDEGQYQCIIIYGVAWDYKYLTLKVKASYRKINTHILKVPETDEVELTCQATGYPLAEVSWPNVSVPANTSHSRTPEGLYQVTSVLRLKPPPGRNFSCVFWNTHVRELTLASIDLQSQMEPRTHPT
- Immunogen:PDCD1LG2 (AAH74766.1, 19 a.a. ~ 220 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant PDCD1LG2.
Type: Primary
Antigen: PDCD1LG2
Clonality: Monoclonal
Clone: 5C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human