Anti-CUEDC2 Mouse Monoclonal Antibody [clone: 2B11]
Catalog # ABNOH00079004-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CUE domain containing 2
- Antigen symbol:CUEDC2
- Clonality:Monoclonal
- Clone:2B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Gene ID:79004
- Antigen synonyms:C10orf66|bA18I14.5
- Amino acid number:1 to 226
- Sequence:MELERIVSAALLAFVQTHLPEADLSGLDEVIFSYVLGVLEDLGPSGPSEENFDMEAFTEMMEAYVPGFAHIPRGTIGDMMQKLSGQLSDARNKENLQPQSSGVQGQVPISPEPLQRPEMLKEETRSSAAAAADTQDEATGAEEELLPGVDVLLEVFPTCSVEQAQWVLAKARGDLEEAVQMLVEGKEEGPAAWEGPNQDLPRRLRGPQKDELKSFILQKYMMVDSA
- Immunogen:CUEDC2 (NP_076945.1, 1 a.a. ~ 226 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant CUEDC2.
Type: Primary
Antigen: CUEDC2
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human