Specifications
- Antibody type:Primary
- Antigen name:MALT1 paracaspase
- Antigen symbol:MALT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Gene ID:10892
- Antigen synonyms:MLT1|IMD12|MLT
- Amino acid number:1 to 813
- Immunogen:MALT1 (NP_776216.1, 1 a.a. ~ 813 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human MALT1 protein.
Type: Primary
Antigen: MALT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human