Order Entry
Export
Orders LinkContactUsLinkComponent
 

Anti-RbcL Rabbit Polyclonal Antibody

50 µg
  AGRIAS03037A
 :   Agrisera
Anti-RbcL Rabbit Polyclonal Antibody
  AGRIAS03037A
 :  Agrisera
Anti-RbcL Rabbit Polyclonal Antibody

50 µg

 
487,00 €
   
1 * 50 µG
 

This antibody is especially suitable for quantifying of Rubisco in plant and algal samples. Rubisco (Ribulose-1,5-bisphosphate carboxylase/oxygenase) catalyzes the rate-limiting step of CO2 fixation in photosynthetic organisms. It is demonstrably homologous from purple bacteria to flowering plants and consists of two protein subunits, each present in 8 copies. In plants and green algae, the large subunit (~55 kDa) is coded by the chloroplast rbcL gene, and the small subunit (15 kDa) is coded by a family of nuclear rbcS genes.,This antibody is especially suitable for quantifying of Rubisco in plant and algal samples. Rubisco (Ribulose-1,5-bisphosphate carboxylase/oxygenase) catalyzes the rate-limiting step of CO2 fixation in photosynthetic organisms. It is demonstrably homologous from purple bacteria to flowering plants and consists of two protein subunits, each present in 8 copies. In plants and green algae, the large subunit (~55 kDa) is coded by the chloroplast rbcL gene, and the small subunit (15 kDa) is coded by a family of nuclear rbcS genes.

Type: Primary
Antigen: RBCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. graveolens, A. thaliana, A. annua, B. sphaerulata, B. sinuspersici, C. raudensis, C. reinhardtii, C. arietinum, C. quitensis kunt bartl, C. paradoxa, C. raciborskii cs-505, Dinoflagellate endosymbionts, E. huxleyi, E. gracilis, extreme acidophilic verrucomicrobial methanotroph methylacidiphilum fumariolicum strain solv, F. mandshurica, F. vesiculosus, Glycine max, G. polyedra, Guzmania hybrid, Heterosigma akashiwo, K. brevis, Moestrup, Chicken, Liquidambar formosana, M. pusila, N. benthamiana, Physcomitrella patens, Porphyra sp., Prochlorococcus sp., Schima superba, Spinacia oleracea, Stanleya pinnata, Symbiodinium sp., Synechococcus pcc 7942, Thalassiosira pseudonana, Thalassiosira punctigera, T. elongatus, T. aestivum, V. vinifera

 
  • Size:
    50 µg
  • Western blot:
    Yes
  • Cat. No.:
    AGRIAS03037A
  • Clonality:
    Polyclonal
  • Reactivity:
    Prochlorococcus sp.|Dinoflagellate endosymbionts|C. arietinum|Thalassiosira punctigera|Chicken|G. polyedra|A. annua|Vitis vinifera|Extreme acidophilic verrucomicrobial methanotroph methylacidiphilum fumariolicum strain solv|C. raudensis|Porphyra sp.|Liquidambar formosana|E. gracilis|C. quitensis kunt bartl|K. brevis|C. raciborskii cs-505|Schima superba|Soy bean|A. graveolens|M. pusila|Stanleya pinnata|Physcomitrella patens|Thermosynechococcus elongatus|Moestrup|Synechococcus sp. pcc 7942|C. paradoxa|B. sphaerulata|Guzmania hybrid|B. sinuspersici|Spinacia oleracea|Thalassiosira pseudonana|T. aestivum|F. mandshurica|F. vesiculosus|E. huxleyi|N. benthamiana|Symbiodinium sp.|C. reinhardtii|A. thaliana|Heterosigma akashiwo
  • Antibody type:
    Primary
  • Immunogen:
    KLH-conjugated synthetic peptide conserved across all known plant, algal and (cyano)bacterial RbcL protein sequences (form I L8S8 and form II L2), including Arabidopsis thaliana AtCg00490, Hordeum vulgare P05698, Oryza sativa P0C510, Chlamydomonas reinhardtii P00877, Synechococcus PCC 7920 A5CKC5
  • Host:
    Rabbit
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    RBCL