Order Entry
Export
ContactUsLinkComponent
 
Anti-ITGA2B Rabbit Polyclonal Antibody
Catalog # BSBTPB9647
Supplier:  Boster Bio
undefined
Anti-ITGA2B Rabbit Polyclonal Antibody
Catalog # BSBTPB9647
Supplier:  Boster Bio

Specifications

  • Antigen name:
    Integrin Alpha 2 Beta
  • Antigen symbol:
    ITGA2B
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    3674
  • Antigen synonyms:
    PPP1R93|GP2B|GTA|CD41B|HPA3|GPIIb|BDPLT2|CD41|BDPLT16|GT
  • UniProtKB:
    P08514
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human ITGA2B (677-711aa EAELAVHLPQGAHYMRALSNVEGFERLICNQKKEN), different from the related mouse sequence by four amino acids.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 μg / vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Integrin alpha-Iib(ITGA2B) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Integrin alpha-IIb is a protein that in humans is encoded by the ITGA2B gene. It is mapped to 17q21.32. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. Alpha chain 2b undergoes post-translational cleavage to yield disulfide-linked light and heavy chains that join with beta 3 to form a fibrinogen receptor expressed in platelets that plays a crucial role in coagulation. Mutations that interfere with this role result in thrombasthenia. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.