Specifications
- Antigen name:endogenous retrovirus group W, member 1
- Antigen symbol:ERVW-1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Gene ID:30816
- Antigen synonyms:HERVW|HERV7Q|ERVWE1|HERV-W-ENV|ENV|ENVW|HERVWENV|HERV-7q
- UniProtKB:Q9UQF0
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ERVW-1(406-435aa YYVNQSGIVTEKVKEIRDRIQRRAEELRNT).
- Purification:Immunogen affinity purified.
- Size:100 μg / vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Syncytin-1(ERVW-1) detection. Tested with WB in Human;Mouse;Rat.
ERVW-1 is also known as ERVWE1. The human ERVWE1 locus is derived from a human endogenous retrovirus-W (HERV-W) provirus located on chromosome 7. This provirus has inactivating mutations in the gag and pol genes, but the envelope glycoprotein gene has been selectively preserved. The product of this gene, syncytin, is expressed in the placental syncytiotrophoblast and is involved in fusion of the cytotrophoblast cells to form the syncytial layer of the placenta. The protein has the characteristics of a typical retroviral envelope protein, including a furin cleavage site that separates the surface (SU) and transmembrane (TM) proteins which form a heterodimer.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.