Capillary columns, Rtx®-50 (fused silica)
Supplier: Restek
Certificates
About this item
General purpose columns for pesticides, herbicides, rosin acids, phthalate esters, sterols. The high thermal stability of Rtx®-50 columns makes dual column analysis possible with common phases such as Rtx®-1 or Rtx®-5MS.
Midpolarity phase; Crossbond® phenyl methyl polysiloxane.
Temperature range: 40 to 320 °C
Similar phases: HP-50+, CP-Sil 24 CB, SPB-50, AT-50, 007-17
Equivalent to USP G3 phase.
Specifications
- Catalog No:
- REST10553
- REST10554
- REST40511
- REST10543
- REST10550
- REST10551
- REST10552
- REST10539
- REST10522
- REST10540
- REST10541
- REST10542
- REST10520
- REST10521
- REST40510
- REST40502
- REST10536
- REST10537
- REST10538
- REST10569
- REST10526
- REST40501
- REST10570
- REST10527
- REST10573
- REST10535
- REST10556
- REST10557
- REST10523
- REST10558
- REST10524
- REST10567
- REST10555
- Temperature limit:
- +40...+280/300 °C
- +40...+280/300 °C
- +40...+300/320 °C
- +40...+270/290 °C
- +40...+280/300 °C
- +40...+280/300 °C
- +40...+260/280 °C
- +40...+290/310 °C
- +40...+280/300 °C
- +40...+270/290 °C
- +40...+290/310 °C
- +40...+290/310 °C
- +40...+300/320 °C
- +40...+300/320 °C
- +40...+300/320 °C
- +40...+310/330 °C
- +40...+290/310 °C
- +40...+270/290 °C
- +40...+290/310 °C
- +40...+270/290 °C
- +40...+300/320 °C
- +40...+310/330 °C
- +40...+250/270 °C
- +40...+300/320 °C
- +40...+250/270 °C
- +40...+290/310 °C
- +40...+280/300 °C
- +40...+280/300 °C
- +40...+300/320 °C
- +40...+260/280 °C
- +40...+300/320 °C
- +40...+250/270 °C
- +40...+260/280 °C
- USP code:
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3
- G3