Orders LinkContactUsLinkComponent
TDS meter, handheld, HI 9034
TDS meter, handheld, HI 9034
Catalog #
TDS meter, handheld, HI 9034
Catalog #
Supplier:  
TDS meter, handheld, HI 9034
Product Details & Documents
HI 9034 measures total dissolved solids (TDS) in three ranges to offer the highest accuracy when performing measurements in diverse applications such as HVAC, waste water treatment and reverse osmosis. All three ranges can be activated at the touch of a button without having to change the probe.
  • Water resistant casing
  • Large dual level LCD is easy to read, even in tough conditions
  • On-screen tutorial messaging
  • BEPS (Battery Error Prevention System) technology alerts the user when low batteries could affect the readings
  • With ATC (automatic temperature compensation)
Delivery information: Supplied with HI 76302W conductivity probe with 1 m cable, calibration screwdriver, battery (1×9 V), rugged carrying case, instruction manual.