Order Entry
Export
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SK3]
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SK3]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SK3]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-0047-T100
  • 35-0047-T500
  • 35-0047-T025
  • Antigen symbol:
  • CD4
  • CD4
  • CD4
  • Antigen name:
  • CD4 molecule
  • CD4 molecule
  • CD4 molecule
  • Antigen synonyms:
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • SK3
  • SK3
  • SK3
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 920
  • 920
  • 920
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications