Order Entry
Export
ContactUsLinkComponent
Anti-DKK2 Rabbit Polyclonal Antibody
Anti-DKK2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9551
Supplier:  Boster Bio
undefined
Anti-DKK2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9551
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dickkopf WNT signaling pathway inhibitor 2
  • Antigen symbol:
    DKK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    27123
  • Antigen synonyms:
    DKK-2
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Dickkopf-related protein 2(DKK2) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: DKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat