Anti-HRPT2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9532
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cell division cycle 73
- Antigen symbol:CDC73
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79577
- Antigen synonyms:FIHP|C1orf28|HRPT2|HYX|HPTJT|HRPT1
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Parafibromin(CDC73) detection. Tested with WB in Human.
Type: Primary
Antigen: HRPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human