Order Entry
Export
ContactUsLinkComponent
Anti-BAG2 Rabbit Polyclonal Antibody
Anti-BAG2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9524
Supplier:  Boster Bio
undefined
Anti-BAG2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9524
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCL2-associated athanogene 2
  • Antigen symbol:
    BAG2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9532
  • Antigen synonyms:
    dJ417I1.2|BAG-2
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for BAG family molecular chaperone regulator 2(BAG2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: BAG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat