Order Entry
Export
ContactUsLinkComponent
Anti-HK1 Polyclonal Antibody Pair
Anti-HK1 Polyclonal Antibody Pair
Catalog # ABNOH00003098-PW2
Supplier:  Abnova
Anti-HK1 Polyclonal Antibody Pair
Catalog # ABNOH00003098-PW2
Supplier:  Abnova

Specifications

  • Antigen name:
    Hexokinase 1
  • Antigen symbol:
    HK1
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3098
  • Antigen synonyms:
    HKD|HK1-tb|HKI|HMSNR|HXK1|HK1-ta|HK1-tc
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse purified Polyclonal Anti-HK1 (300 µg); 2. Antibody pair for Western Blot: Rabbit purified Polyclonal Anti-HK1 (50 µg)

Type:
Antigen: HK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human