Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-RAB3B Antibody Pair

Anti-RAB3B Antibody Pair

1 Set
Catalog # ABNOH00005865-PW1
Supplier:   Abnova
Anti-RAB3B Antibody Pair
Catalog # ABNOH00005865-PW1
Supplier:  Abnova
Anti-RAB3B Antibody Pair
Anti-RAB3B Antibody Pair

1 Set

Price
885,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-RAB3B (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-RAB3B (50 µl)

Type:
Antigen: RAB3B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human

Specifications
  • Size:
    1 Set
  • Western blot:
    Yes
  • Cat. No.:
    ABNOH00005865-PW1
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Gene ID:
    5865
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Host:
    Multiple
  • Antigen symbol:
    RAB3B
  • Antigen name:
    Ras-related protein Rab-3B