Order Entry
Export
ContactUsLinkComponent
Anti-CBR3 Polyclonal Antibody Pair
Anti-CBR3 Polyclonal Antibody Pair
Catalog # ABNOH00000874-AP21
Supplier:  Abnova
Anti-CBR3 Polyclonal Antibody Pair
Catalog # ABNOH00000874-AP21
Supplier:  Abnova

Specifications

  • Antigen name:
    Carbonyl Reductase 3
  • Antigen symbol:
    CBR3
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    874
  • Antigen synonyms:
    hCBR3|SDR21C2|HEL-S-25
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human CBR3.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CBR3 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-CBR3 (20 µg);

Type:
Antigen: CBR3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human