Anti-ULK3 Rabbit Polyclonal Antibody
Catalog # BSBTPB9519
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Unc-51 like kinase 3
- Antigen symbol:ULK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:25989
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase ULK3(ULK3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: ULK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat