Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-SLC19A1 Rabbit Polyclonal Antibody

Anti-SLC19A1 Rabbit Polyclonal Antibody

100 µg
Catalog # BSBTPB9504
Supplier:   Boster Bio
Anti-SLC19A1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9504
Supplier:  Boster Bio
Anti-SLC19A1 Rabbit Polyclonal Antibody
Anti-SLC19A1 Rabbit Polyclonal Antibody

100 µg

Price
546,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Rabbit IgG polyclonal antibody for Folate transporter 1(SLC19A1) detection. Tested with WB in Human.

Type: Primary
Antigen: SLC19A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Documents

Specifications
  • Size:
    100 µg
  • Western blot:
    Yes
  • Cat. No.:
    BSBTPB9504
  • Clonality:
    Polyclonal
  • Reactivity:
    Human
  • Antibody type:
    Primary
  • Gene ID:
     6573
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    Solute carrier family 19, member 1
  • Cross adsorption:
    No