Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-SCARB1 Rabbit Polyclonal Antibody

Anti-SCARB1 Rabbit Polyclonal Antibody

100 µg
Catalog # BSBTPB9502
Supplier:   Boster Bio
Anti-SCARB1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9502
Supplier:  Boster Bio
Anti-SCARB1 Rabbit Polyclonal Antibody
Anti-SCARB1 Rabbit Polyclonal Antibody

100 µg

Price
546,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Rabbit IgG polyclonal antibody for Scavenger receptor class B member 1(SCARB1) detection. Tested with WB in Mouse;Rat.

Type: Primary
Antigen: SCARB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Documents

Specifications
  • Size:
    100 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Antibody type:
    Primary
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Antigen name:
    Scavenger receptor class B, member 1
  • Cat. No.:
    BSBTPB9502
  • Reactivity:
    Rat|Mouse
  • Gene ID:
    20778
  • Antigen synonyms:
    SR-BI|mSR-BI|SR-B1|CD36|Cla-1|Hdlq1|Hlb398|SRBI|Srb1|D5Ertd460e|Cd36l1|Cla1|AI120173
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    SCARB1