Anti-ALDH2 Rabbit Polyclonal Antibody
100 µg
Catalog # BSBTPB9472
Supplier: Boster Bio
Jump to:
Product Details & Documents
Rabbit IgG polyclonal antibody for Aldehyde dehydrogenase, mitochondrial(ALDH2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: ALDH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
PB9472
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Cross adsorption:No
- Antigen name:aldehyde dehydrogenase 2 family (mitochondrial)
- Cat. No.:BSBTPB9472
- Reactivity:Human|Rat|Mouse
- Gene ID:217
- Antigen synonyms:ALDM|ALDH-E2|ALDHI
- Conjugation:Unconjugated
- Antigen symbol:ALDH2



