Anti-RAGE Rabbit Polyclonal Antibody
BSBTPB9469
- Antibody type:Primary
- Antigen name:advanced glycosylation end product-specific receptor
- Antigen symbol:AGER
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:177
- Antigen synonyms:RAGE
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Advanced glycosylation end product-specific receptor (AGER) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: RAGE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat